Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mPEG, Mouse anti-All, Clone: 15-2b, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $530.40
Specifications
| Antigen | mPEG |
|---|---|
| Clone | 15-2b |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABS200225
|
MilliporeSigma
MABS200225UG |
25 μg |
Each for $192.34
|
|
|||||
|
MABS2002100
|
MilliporeSigma
MABS2002100UG |
100 μg |
Each for $530.40
|
|
|||||
Description
Anti-mPEG, clone 15-2b, Cat. No. MABS2002, is a mouse monoclonal antibody that detects methoxy-PEG (mPEG)-conjugated proteins and has been tested for use in ELISA.
Polyethylene glycols (PEG) are non-toxic, uncharged, hydrophilic long chain polymers with a big exclusion volume in aqueous solutions. The covalent attachment of PEG is commonly employed to modify proteins and drugs that can increase their bioavailability and reduce immunogenicity and proteolytic degradation. The monomethoxylated form of polyethylene glycol (mPEG) has been widely employed in drug modifications. mPEG modification reduces nonspecific binding and prevents recognition by the reticuloendothelial system. This monoclonal antibody (clone 15-2b) specifically binds to the terminal methoxy group, but not the backbone of PEG. It displays high affinity for the terminal methoxy group and can be easily used as a detection antibody for the quantification of mPEG by ELISA. It is also suitable for detection of PEGylated liposomes and nanoparticles. The mPEG cell-based competition ELISA can detect lower molecular-weight (< 2 kDa) mPEG molecules even at nanomolar concentration and the assay is not affected by the presence of mouse or human serum. In addition, mPEG cell-based ELISA can be useful in determining the pharmacokinetics of a mPEG molecule an animal without the need for any radiolabeling.Specifications
| mPEG | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Signaling | |
| Purified mouse monoclonal antibody IgG2b in PBS with 0.04% sodium azide. | |
| Methoxy PEG covalently conjugated to E.coli beta-glucuronidase. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
| 15-2b | |
| Monoclonal | |
| Purified | |
| RUO | |
| All species | |
| Methoxy polyethylene glycol | |
| IgG2b κ | |
| Protein A purified | |
| Clone 15-2b specifically detects proteins conjugated with methoxy polyethylene glycol (mPEG). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title