Learn More
Invitrogen™ MPEG1 Polyclonal Antibody

Description
PA1-29037 detects MPEG1 from mouse, rat samples.
Specifications
Specifications
| Antigen | MPEG1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Mpeg1 |
| Gene Accession No. | A1L314, Q9WV57 |
| Gene Alias | macrophage expressed 1; macrophage expressed gene 1; macrophage gene 1 protein; Macrophage-expressed gene 1 protein; MGC150002; MGC150003; MPEG1; MPG1; Mpg-1; MPS1; P-2; perforin 2; Perforin-2; Protein MPS1 |
| Gene Symbols | Mpeg1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.