Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPHOSPH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259106 Supplier Novus Biologicals Supplier No. NBP259106

Rabbit Polyclonal Antibody

MPHOSPH1 Polyclonal specifically detects MPHOSPH1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen MPHOSPH1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias Cancer/testis antigen 90, CT90MPP-1, DKFZp434B0435, EC 6.3.5.5, kinesin family member 20B, Kinesin-related motor interacting with PIN1, KRMP1kinesin-like protein KIF20B, M-phase phosphoprotein 1mitotic kinesin-like protein, MPHOSPH1EC 2.1.1, MPP1DKFZp434P0810
Gene Symbols KIF20B
Host Species Rabbit
Immunogen Maps to a region between 825 and 875
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9585
Target Species Human
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.