Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mPlum Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB151631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151631 Supplier Novus Biologicals Supplier No. NBP311833JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mPlum Polyclonal antibody specifically detects mPlum in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen mPlum
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Fluorescent protein plum, mPlum Fluorescent Protein, Red Fluorescent Protein, RFP
Host Species Rabbit
Immunogen mPlum from Discosoma sp.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.