Learn More
Description
Specifications
Specifications
| Antigen | MPP2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Discs large homolog 2, DKFZp686A06252, DKFZp686J2189, DKFZp761D0712, DLG2discs large, homolog 2, MAGUK p55 subfamily member 2, membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2), Protein MPP2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human MPP2 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
