Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB048469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048469 Supplier Novus Biologicals Supplier No. NBP297850C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MPP2 Polyclonal specifically detects MPP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MPP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Discs large homolog 2, DKFZp686A06252, DKFZp686J2189, DKFZp761D0712, DLG2discs large, homolog 2, MAGUK p55 subfamily member 2, membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2), Protein MPP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MPP2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4355
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.