Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582919
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA582919 100 μL
1 options

Catalog No. PIPA582919

Supplier: Invitrogen™ PA582919

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRAMNRAALE SGISTKQLTE ADLRRTVEES SRIQRGYGHY FDLCLVNSNL ERTFRELQT Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP2 (membrane protein, palmitoylated 2), also known as MAGUK p55 subfamily member 2, discs large homolog 2 or DLG2, is a 576 amino acid protein belonging to the MAGUK family that exists as three alternatively spliced isoforms. MPP2 contains one guanylate kinase-like domain, a PDZ (DHR) domain, two L27 domains and a single SH3 domain. The gene encoding MPP2 maps to the same segment of human chromosome 17 as MPP3, with whom MMP2 likely shares similar function and common structural organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mpp2
Gene Accession No. Q14168
Gene Alias D11Bwg0652e; discs large homolog 2; discs large, homolog 2; Dlg2; Dlgh2; MAGUK p55 subfamily member 2; membrane palmitoylated protein 2; membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2); Mpp2; p55 subfamily member of MAGUK family; palmitoylated membrane protein 2; Pals4; Protein MPP2; rCG_34691
Gene Symbols Mpp2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPP2. Recombinant protein control fragment (Product #RP-93215).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.