Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP3 Antibody (OTI8F1), Novus Biologicals™
SDP

Catalog No. NBP202659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202659 Supplier Novus Biologicals Supplier No. NBP202659
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MPP3 Monoclonal specifically detects MPP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MPP3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI8F1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Gene Alias Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3
Gene Symbols MPP3
Host Species Mouse
Immunogen Full length human recombinant protein of human MPP3 (NP_001923) produced in HEK293T cell.
Molecular Weight of Antigen 66 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4356
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.