Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mpp8 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291167 Supplier Novus Biologicals Supplier No. NBP171809F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mpp8 Polyclonal antibody specifically detects mpp8 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen mpp8
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ35237, HSMPP8, M-phase phosphoprotein 8, M-phase phosphoprotein, mpp, M-phase phosphoprotein, mpp8, MPP8, TWA3, Two hybrid-associated protein 3 with RanBPM
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 200 and 250 of human M-phase Phosphoprotein 8 using the numbering given in entry NP_059990.2 (GeneID 54737).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54737
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.