Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPPED2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | MPPED2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MPPED2 Polyclonal specifically detects MPPED2 in Human samples. It is validated for Western Blot.Specifications
| MPPED2 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| 239FBchromosome 11 open reading frame 8, C11orf8dJ1024C24.1, D11S302E, dJ873F21.1, EC 3.1, FAM1B, Fetal brain protein 239, metallophosphoesterase domain containing 2, Metallophosphoesterase domain-containing protein 2, metallophosphoesterase MPPED2 | |
| MPPED2 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001575 | |
| 744 | |
| Synthetic peptide directed towards the N terminal of human MPPED2. Peptide sequence RFQPPHVHMVDPIPYDTPKPAGHTRFVCISDTHSRTDGIQMPYGDILLHT. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title