Learn More
Invitrogen™ MPS1 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Caco-2 whole cell, human RT4 whole cell, human Jurkat whole cell, rat PC-12 whole cell, rat RH35 whole cell, mouse RAW2647 whole cell, mouse SP2/0 whole cell. IHC: human intestinal cancer tissue, mouse brain cancer tissue, rat brain cancer tissue. ICC/IF: U20S cell. Flow: HeLa cell.
Specifications
Specifications
| Antigen | MPS1 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide |
| Gene | RPS27 |
| Gene Accession No. | P42677, Q6ZWU9, Q71TY3 |
| Gene Alias | 3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 |
| Gene Symbols | RPS27 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.