Learn More
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 117871AP150UL
Description
This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.
Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.Specifications
| MPZL2 | |
| Polyclonal | |
| Unconjugated | |
| MPZL2 | |
| Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10205, 14012, 300679 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60487, O70255 | |
| MPZL2 | |
| MPZL2 Fusion Protein Ag2419 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.