Learn More
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | MPZL2 |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7454
![]()
|
Proteintech Group Inc
117871AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.
Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.Specifications
| MPZL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60487, O70255 | |
| 10205, 14012, 300679 | |
| MPZL2 Fusion Protein Ag2419 | |
| Primary | |
| -20°C | |
| MPZL2 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2 | |
| MPZL2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.