Learn More
MPZL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MPZL3 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4940
![]() ![]() |
Proteintech Group Inc
255131AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This polyclonal antibody raised against 32-235 aa of human MPZL3 reveals bands of 28 kDa and 70 kDa, which could be endogenous MPZL3 of unmodified form and post-translational modified form, respectively.
MPZL3 mediates homophilic cell-cell adhesion.Specifications
MPZL3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q3V3F6, Q6UWV2 | |
196264, 319742 | |
MPZL3 Fusion Protein Ag21905 | |
Primary | |
-20°C | |
MPZL3 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MPZL3, myelin protein zero like 3 | |
Mpzl3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.