Learn More
Description
Specifications
Specifications
| Antigen | MR1 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 155-205 on human Major Histocompatibility Complex Class I related gene protein, isoform 1/ MHC I. |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
