Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MR1, Mouse anti-Rat, Human, Mouse, Bovine, Clone: 26.5, Millipore Sigma™

Catalog No. MABF207525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF207525 25 μL
MABF2075100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF207525 Supplier MilliporeSigma Supplier No. MABF207525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MR1, clone 26.5, Cat. No. MABF2075, is a mouse monoclonal antibody that detects MHC class I-related gene protein and has been tested for use in Flow Cytometry, Function Analysis, and Immunoprecipitation.

Major histocompatibility complex class I-related gene protein (UniProt: Q95460; also known as MHC class I-related gene protein, Class I histocompatibility antigen-like protein, MR1) is encoded by the MR1 gene (Gene ID: 3140) in human. MR1 is a single-pass membrane protein that is ubiquitous in its distribution and can heterodimerize with B2M. It serves as an antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. It is involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain known as mucosal-associated invariant T-cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and may be involved in monitoring commensal flora or serve as a distress signal. MR1-restricted T cells can discriminate between microbes in a TCR-dependent manner. However, these cells are shown to be severely reduced in circulation in subjects with chronic HIV-1 infection. MR1 is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off in the mature form. The mature form has an extracellular domain (aa 23-302), a transmembrane domain (aa-303-323), and a short cytoplasmic tail (aa 324-341). Its ligand binding region is localized to amino acids 23-201. Five different isoforms of MR1 have been described that are produced by alternative splicing. (Ref.: Young, MH et al., (2013). PLOS one 8(1); e53789; Leeansyah, E., et al. (2013). Blood. 121(7); 1124-1135).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MR1
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone 26.5
Conjugate Unconjugated
Dilution Immunoprecipitation Analysis: A representative lot immunoprecipitated MR1 in Immunoprecipitation applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93).Flow Cytometry Analysis: A representative lot detected MR1 in flow cytometry applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93; Gold, M.C., et. al. (2010). PLoS Biol. 8(6):e1000407; Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5). Inhibits Activity/Function Analysis: A representative lot inhibited the activation of mucosal-associated invariant T (MAIT) cells by MR1 protein. (Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5).
Formulation Purified mouse monoclonal antibody IgG2a in PBS without azide.
Gene Alias Major histocompatibility complex class I-related gene protein;MHC class I-related gene protein;Class I histocompatibility antigen-like protein
Gene Symbols MR1
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 280 amino acids from the extracellular domain of human MHC class I-related gene protein.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001522.1
Test Specificity Clone 26.5 specifically detects Major histocompatibility complex class I-related gene protein (MR1). It targets an epitope with in the extracellular domain.
Target Species Rat, Human, Mouse, Bovine
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.