Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MR1, Mouse anti-Rat, Human, Mouse, Bovine, Clone: 26.5, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $482.80
Specifications
Antigen | MR1 |
---|---|
Clone | 26.5 |
Dilution | Immunoprecipitation Analysis: A representative lot immunoprecipitated MR1 in Immunoprecipitation applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93).Flow Cytometry Analysis: A representative lot detected MR1 in flow cytometry applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93; Gold, M.C., et. al. (2010). PLoS Biol. 8(6):e1000407; Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5). Inhibits Activity/Function Analysis: A representative lot inhibited the activation of mucosal-associated invariant T (MAIT) cells by MR1 protein. (Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5). |
Applications | Flow Cytometry, Functional Assay, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF207525
|
MilliporeSigma
MABF207525UL |
25 μL |
Each for $192.34
|
|
|||||
MABF2075100
|
MilliporeSigma
MABF2075100UL |
100 μL |
Each for $482.80
|
|
|||||
Description
Anti-MR1, clone 26.5, Cat. No. MABF2075, is a mouse monoclonal antibody that detects MHC class I-related gene protein and has been tested for use in Flow Cytometry, Function Analysis, and Immunoprecipitation.
Major histocompatibility complex class I-related gene protein (UniProt: Q95460; also known as MHC class I-related gene protein, Class I histocompatibility antigen-like protein, MR1) is encoded by the MR1 gene (Gene ID: 3140) in human. MR1 is a single-pass membrane protein that is ubiquitous in its distribution and can heterodimerize with B2M. It serves as an antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. It is involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain known as mucosal-associated invariant T-cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and may be involved in monitoring commensal flora or serve as a distress signal. MR1-restricted T cells can discriminate between microbes in a TCR-dependent manner. However, these cells are shown to be severely reduced in circulation in subjects with chronic HIV-1 infection. MR1 is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off in the mature form. The mature form has an extracellular domain (aa 23-302), a transmembrane domain (aa-303-323), and a short cytoplasmic tail (aa 324-341). Its ligand binding region is localized to amino acids 23-201. Five different isoforms of MR1 have been described that are produced by alternative splicing. (Ref.: Young, MH et al., (2013). PLOS one 8(1); e53789; Leeansyah, E., et al. (2013). Blood. 121(7); 1124-1135).Specifications
MR1 | |
Immunoprecipitation Analysis: A representative lot immunoprecipitated MR1 in Immunoprecipitation applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93).Flow Cytometry Analysis: A representative lot detected MR1 in flow cytometry applications (Huang, S., et. al. (2005). J Biol Chem. 280(22):21183-93; Gold, M.C., et. al. (2010). PLoS Biol. 8(6):e1000407; Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5). Inhibits Activity/Function Analysis: A representative lot inhibited the activation of mucosal-associated invariant T (MAIT) cells by MR1 protein. (Huang, S., et. al. (2009). Proc Natl Acad Sci U S A. 106(20):8290-5). | |
Monoclonal | |
Purified | |
RUO | |
Rat, Human, Mouse, Bovine | |
Major histocompatibility complex class I-related gene protein;MHC class I-related gene protein;Class I histocompatibility antigen-like protein | |
MR1 | |
IgG2a κ | |
Protein G purified | |
Clone 26.5 specifically detects Major histocompatibility complex class I-related gene protein (MR1). It targets an epitope with in the extracellular domain. |
26.5 | |
Flow Cytometry, Functional Assay, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal antibody IgG2a in PBS without azide. | |
NP_001522.1 | |
GST-tagged recombinant fragment corresponding to 280 amino acids from the extracellular domain of human MHC class I-related gene protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title