Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mre11 Antibody, Novus Biologicals™
SDP

Catalog No. NB100142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100142 0.05 mL
NB100142SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100142 Supplier Novus Biologicals Supplier No. NB100142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mre11 Polyclonal specifically detects Mre11 in Chicken, Hamster, Human, Mouse, Rat, Virus samples. It is validated for Chemotaxis, ChIP Assay, ChIP Cytometry, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, KnockDown, Proximity Ligation Assay, Simple Western, Western Blot.

Specifications

Antigen Mre11
Applications Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, KnockDown, Proximity Ligation Assay, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Simple Western 1:250, Flow Cytometry, ELISA reported in scientific literature (PMID 16788144), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200. Use reported in scientific literature (PMID 26774475), Immunoprecipitation 3 uL, Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 21279473), Immunohistochemistry-Frozen reported in scientific literature (PMID 24349281), Immunoblotting reported in scientific literature (PMID 28115467), Proximity Ligation Assay reported in scientific literature (PMID 32780723), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (VanCevska et al), KnockDown Validated
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. P49959
Gene Alias ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B
Gene Symbols MRE11
Host Species Rabbit
Immunogen Mre11 Antibody is made to a full length human Mre11 protein. [Uniprot: P49959]
Molecular Weight of Antigen 81 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 4361
Target Species Chicken, Mouse, Hamster, Human, Mouse, Rat, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.