Learn More
Description
Specifications
Specifications
| Antigen | Mre11 |
| Applications | Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, KnockDown, Proximity Ligation Assay, Simple Western, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000, Simple Western 1:250, Flow Cytometry, ELISA reported in scientific literature (PMID 16788144), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200. Use reported in scientific literature (PMID 26774475), Immunoprecipitation 3 uL, Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 21279473), Immunohistochemistry-Frozen reported in scientific literature (PMID 24349281), Immunoblotting reported in scientific literature (PMID 28115467), Proximity Ligation Assay reported in scientific literature (PMID 32780723), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (VanCevska et al), KnockDown Validated |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Accession No. | P49959 |
| Gene Alias | ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B |
| Gene Symbols | MRE11 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
