Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRGPRF Antibody (916219), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346130 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346130 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346130 Supplier Novus Biologicals Supplier No. FAB8396MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRGPRF Monoclonal specifically detects MRGPRF in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen MRGPRF
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 916219
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MRGPRF, Met1-Ser343, Accession # NP_659452
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 116535
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.