Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRGX2 Antibody (477533), FITC, Novus Biologicals™
SDP

Catalog No. NB369180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369180 Supplier Novus Biologicals Supplier No. FAB4727F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRGX2 Monoclonal specifically detects MRGX2 in Human samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining).

Specifications

Antigen MRGX2
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 477533
Conjugate FITC
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGX2G protein-coupled receptor MRGX2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MRGX2, Met1-Val330, Accession # Q96LB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 117194
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.