Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRGX2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018024 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018024 0.025 mL
NB11075035 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018024 Supplier Novus Biologicals Supplier No. NB110750350.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

MRGX2 Polyclonal specifically detects MRGX2 in Human, Mouse, Hamster (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Single Cell Western, Knockdown Validated.

Specifications

Antigen MRGX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 : 200 - 1 : 2000, Immunohistochemistry 1 : 200 - 1 : 2000
Formulation Whole antisera with No Preservative
Gene Accession No. Q96LB1
Gene Alias MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGX2G protein-coupled receptor MRGX2
Gene Symbols MRGPRX2
Host Species Rabbit
Immunogen A synthetic peptide from the 4th cytoplasmic loop of human Mas-related G-protein coupled receptor member X2 (MRGPRX2) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 117194
Test Specificity MRGPRX2.
Reconstitution Reconstitute with 0.1 ml sterilized water.
Target Species Hamster (Negative), Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.