Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRK 016
GREENER_CHOICE

Catalog No. p-4817038
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α5-selective GABAA inverse agonist

GABAA receptor inverse agonist selective for the α5-subtype (EC50 = 3 nM). Exhibits affinity at benzodiazepine binding site of recombinant human GABAA receptors (Ki values are 0.77 nM, 0.83 nM, 0.85 nM and 1.4 nM for α3-, α1-, α2-, and α5-containing respectively). Increases long-term potentiation (LTP) in mouse hippocampal slices. Exhibits no anxiogenic or proconvulsant activity.

Chemical Identifiers

CAS 783331-24-8
Molecular Formula C17H20N8O2

Specifications

Quantity 10 mg
Formula Weight 368.39
Purity >98%
Molecular Weight (g/mol) 368.401
Chemical Name or Material MRK 016
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.