Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRLC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254719 Supplier Novus Biologicals Supplier No. NBP254719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRLC2 Polyclonal specifically detects MRLC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MRLC2
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias MLC-2, MLC20, MLC-2A, MLC-B, MRLC2SHUJUN-1, MYLC2B, myosin regulatory light chain 12B, myosin regulatory light chain 2, Myosin regulatory light chain 20 kDa, Myosin regulatory light chain 2-B, smooth muscle isoform, Myosin regulatory light chain MRLC2, myosin, light chain 12B, regulatory
Gene Symbols MYL12B
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:YREAPIDKKGNFNYIEFTRILKHGAKDKDD
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103910
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.