Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MROH2B Antibody, Novus Biologicals™
SDP

Catalog No. NBP232466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232466 0.1 mL
NB397496 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP232466 Supplier Novus Biologicals Supplier No. NBP232466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MROH2B Polyclonal specifically detects MROH2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MROH2B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q7Z745
Gene Alias HEAT Repeat Family Member 7B2, HEAT Repeat Family Member B2, HEAT Repeat-Containing Protein 7B2, HEATR7B2, Maestro Heat-Like Repeat Family Member 2B, Maestro Heat-Like Repeat-Containing Protein Family Member 2B
Gene Symbols MROH2B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DKEHIQFLYERSMDALGKLLKTMMWDNVNAEDCQEMFNLLQMWLVSQKEWERERAFQITAKVLTNDIEAPENFKIGSLLGLLAPHSCDTLPTI
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133558
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.