Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Mouse anti-Human, Clone: MRP1/1344, Novus Biologicals™
SDP

Catalog No. NB122140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122140 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122140 Supplier Novus Biologicals Supplier No. NBP30733320UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRP1 Monoclonal specifically detects MRP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MRP1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MRP1/1344
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Host Species Mouse
Immunogen Recombinant full-length human MRP1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4363
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.