Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP2 Antibody (M2III-6), Novus Biologicals™
SDP

Catalog No. NBP142349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142349 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142349 Supplier Novus Biologicals Supplier No. NBP142349
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRP2 Monoclonal specifically detects MRP2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MRP2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M2III-6
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:20-1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen 1:20
Gene Accession No. Q92887
Gene Alias ATP-binding cassette, sub-family C (CFTR/MRP), member 2, Canalicular multidrug resistance protein, canalicular multispecific organic anion transporter 1, CMOAT1, CMOATABC30, CMRP, DJS, KIAA1010, MRP2ATP-binding cassette sub-family C member 2, Multidrug resistance-associated protein 2
Gene Symbols ABCC2
Host Species Mouse
Immunogen The antibody reacts with the MRP2 antigen.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1244
Test Specificity M2III-6 reacts with an internal epitope of cMOAT/MRP2, a 170-180 kD transmembrane protein known as the canalicular multi-organic anion transporter, absent in patients with the Dubin-Johnson syndrome, an autosomal recessive liver disorder characterized by chronic conjugated hyperbilirubinemia. cMOAT/MRP2 is closely related to the multidrug resistance related protein MRP, and cMOAT/MRP2 overexpression has been observed in a subset of cisplatin resistant cell lines. M2III-6 was raised against a bacterial fusion protein of cMOAB/MRP2, containing the 202-amino acid COOH terminal end of the protein. M2III-6 did not cross react with the human MDR1, MRP1, MRP3 and MRP5 gene products.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.