Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP3 Antibody (ABCC3/2971), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB010754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010754 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010754 Supplier Novus Biologicals Supplier No. NBP279837AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRP3 Monoclonal specifically detects MRP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MRP3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ABCC3/2971
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ABC31, ATP-binding cassette sub-family C member 3, ATP-binding cassette, sub-family C (CFTR/MRP), member 3, canalicular multispecific organic anion transporter 2, canicular multispecific organic anion transporter, cMOAT2, EC 3.6.3, EC 3.6.3.44, EC 6.3.2.4, MLP2DKFZp686E22157, MRP3MOAT-DEST90757, multidrug resistance associated protein, Multidrug resistance-associated protein 3, Multi-specific organic anion transporter D
Gene Symbols ABCC3
Host Species Mouse
Immunogen Recombinant fragment (around aa 815-957) of human MRP3 protein (exact sequence is proprietary) (Uniprot: O15438)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.