Learn More
Description
Specifications
Specifications
| Antigen | MRP3 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | M3II-21 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:20-1:50, Simple Western, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunohistochemistry-Frozen 1:20 |
| Formulation | 1 ml serum-free tissue culture supernatant with 0.25 mg/ml immunoglobulin and 0.7% BSA with 0.1% Sodium Azide |
| Gene Alias | ABC31, ATP-binding cassette sub-family C member 3, ATP-binding cassette, sub-family C (CFTR/MRP), member 3, canalicular multispecific organic anion transporter 2, canicular multispecific organic anion transporter, cMOAT2, EC 3.6.3, EC 3.6.3.44, EC 6.3.2.4, MLP2DKFZp686E22157, MRP3MOAT-DEST90757, multidrug resistance associated protein, Multidrug resistance-associated protein 3, Multi-specific organic anion transporter D |
| Gene Symbols | ABCC3 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
