Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | MRP6 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118970
|
Invitrogen™
PA5118970 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5122298
|
Invitrogen™
PA5122298 |
20 μL |
Each for $237.00
|
|
|||||
Description
MRP6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene.Specifications
MRP6 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
ABCC6 | |
ABC34; Abcc1b; ABCC6; Anthracycline resistance-associated protein; ARA; ATP binding cassette subfamily C member 6; ATP-binding cassette sub-family C member 6; ATP-binding cassette, subfamily C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; DCC; dyscalc; Dyscalc1; EST349056; GACI2; liver multidrug resistance-associated protein 6; Mlp1; MLP-1; MOATE; MOAT-E; Mrp6; MRP-like protein 1; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6; multi-specific organic anion transporter E; PXE; PXE1; URG7; URG7 protein; multidrug resistance-associated protein 6 | |
ABCC6 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O88269 | |
81642 | |
Recombinant protein ATP Binding Cassette Transporter C6. The antigen corresponds to amino acid range 1260-1502 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title