Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL12 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB097238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097238 Supplier Novus Biologicals Supplier No. NBP297472AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL12 Polyclonal specifically detects MRPL12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MRPL12
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias 5c5-2, FLJ60124, L12mt, MGC8610, mitochondrial ribosomal protein L12, MRP-L12, MRP-L31/34, MRPL7, MRPL7/L12, RPML1239S ribosomal protein L12, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPL12 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6182
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.