Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL13 Antibody (OTI6A11), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287821 Supplier Novus Biologicals Supplier No. NBP272780APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRPL13 Monoclonal antibody specifically detects MRPL13 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MRPL13
Applications Western Blot
Classification Monoclonal
Clone OTI6A11
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias 39S ribosomal protein L13, mitochondrial, L13, L13A, L13mtRPML13, mitochondrial ribosomal protein L13, MRP-L13, RPL13
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPL13 (NP_054797) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 28998
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.