Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL15 Antibody (OTI7A3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272781G missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBP272781G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. NBP272781G Fournisseur Novus Biologicals Code fournisseur NBP272781G
Ajouter au panier
Ajouter au panier

Mouse Monoclonal Antibody

MRPL15 Monoclonal specifically detects MRPL15 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Spécifications

Antigen MRPL15
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7A3
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias L15mtHSPC145, mitochondrial ribosomal protein L15, MRP-L1539S ribosomal protein L15, mitochondrial, MRP-L7, RPML7
Gene Symbols MRPL15
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 28-296 of human MRPL15 (NP_054894) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 29088.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.