Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL15 Antibody (OTI7A3), PE, Novus Biologicals™
SDP

Catalog No. NB287815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287815 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287815 Supplier Novus Biologicals Supplier No. NBP272781PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRPL15 Monoclonal antibody specifically detects MRPL15 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen MRPL15
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7A3
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias L15mtHSPC145, mitochondrial ribosomal protein L15, MRP-L1539S ribosomal protein L15, mitochondrial, MRP-L7, RPML7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 28-296 of human MRPL15(NP_054894) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 29088
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.