Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL17 Antibody, Novus Biologicals™
SDP

Catalog No. NB173686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173686 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173686 Supplier Novus Biologicals Supplier No. NBP318407
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL17 Polyclonal antibody specifically detects MRPL17 in Human samples. It is validated for Western Blot

Specifications

Antigen MRPL17
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias L17mt, LIP2, LYST-interacting protein 2, LYST-interacting protein LIP2, mitochondrial ribosomal protein L17, MRP-L17, MRP-L26, RPL17L, RPML26,39S ribosomal protein L17, mitochondrial
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 25 and 75..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 63875
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.