Learn More
Description
Specifications
Specifications
| Antigen | MRPL19 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | KIAA0104L19mt, L15mt, MGC20675, mitochondrial, mitochondrial ribosomal protein L19, MRPL1539S ribosomal protein L19, mitochondrial, MRP-L15MRP-L19, RLX1, RPML15 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human MRPL19 fragment. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
