Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL19 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB102176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102176 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102176 Supplier Novus Biologicals Supplier No. NBP297473IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL19 Polyclonal specifically detects MRPL19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MRPL19
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias KIAA0104L19mt, L15mt, MGC20675, mitochondrial, mitochondrial ribosomal protein L19, MRPL1539S ribosomal protein L19, mitochondrial, MRP-L15MRP-L19, RLX1, RPML15
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPL19 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9801
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.