Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL34 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB332115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332115 Supplier Novus Biologicals Supplier No. NBP297371MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL34 Polyclonal antibody specifically detects MRPL34 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MRPL34
Applications Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias L34mtMGC24974, MGC2633, mitochondrial ribosomal protein L34, MRP-L34,39S ribosomal protein L34, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPL34 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 64981
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.