Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS11 Antibody (OTI2E9), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272782A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272782A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272782A405 Supplier Novus Biologicals Supplier No. NBP272782AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRPS11 Monoclonal specifically detects MRPS11 in Human samples. It is validated for Western Blot.

Specifications

Antigen MRPS11
Applications Western Blot
Classification Monoclonal
Clone OTI2E9
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 28S ribosomal protein S11, mitochondrial, Cervical cancer proto-oncogene 2 protein, FLJ22512, FLJ23406, HCC-2S11mt, mitochondrial ribosomal protein S11, MRP-S11, RPMS11
Gene Symbols MRPS11
Host Species Mouse
Immunogen Full length recombinant protein of human MRPS11 (NP_073750) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64963.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.