Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS25 Antibody (38E7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP242628A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP242628A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP242628A32 Supplier Novus Biologicals Supplier No. NBP242628AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRPS25 Monoclonal specifically detects MRPS25 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MRPS25
Applications Western Blot, ELISA
Classification Monoclonal
Clone 380000000
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ00023, mitochondrial 28S ribosomal protein S25, mitochondrial ribosomal protein S25, MRP-S2528S ribosomal protein S25, mitochondrial, RPMS25DKFZp313H0817, S25mt
Gene Symbols MRPS25
Host Species Mouse
Immunogen Recombinant human MRPS25 (1-173aa) purified from E. coli
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 64432
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.