Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS26 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB331577 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331577 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331577 Supplier Novus Biologicals Supplier No. NBP297474PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPS26 Polyclonal antibody specifically detects MRPS26 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen MRPS26
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C20orf193, dJ534B8.3, GI008,28S ribosomal protein S13, mitochondrial, mitochondrial ribosomal protein S26, MRP-S13MRPS13, MRP-S26chromosome 20 open reading frame 193, NY-BR-87, RPMS1328S ribosomal protein S26, mitochondrial, S13mt, S26mt, serologically defined breast cancer antigen NY-BR-87
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPS26 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 64949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.