Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS27 Antibody (OTI1A2), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272784PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272784PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272784PEC5 Supplier Novus Biologicals Supplier No. NBP272784PECY55

Mouse Monoclonal Antibody

MRPS27 Monoclonal specifically detects MRPS27 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MRPS27
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1A2
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ21764, KIAA0264FLJ23348, mitochondrial, mitochondrial ribosomal protein S27, MRP-S27
Gene Symbols MRPS27
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 (NP_055899) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23107.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.