Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS35 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB054286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054286 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054286 Supplier Novus Biologicals Supplier No. NBP298524F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPS35 Polyclonal specifically detects MRPS35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MRPS35
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 28S ribosomal protein S28, mitochondrial, DKFZp762P093, MDS023,28S ribosomal protein S35, mitochondrial, MGC104278, mitochondrial ribosomal protein S28, mitochondrial ribosomal protein S35, MRP-S28, MRPS28S35mt, MRP-S35, S28mt
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPS35 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60488
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.