Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS7 Antibody (OTI5E11), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272789A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272789A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272789A48 Supplier Novus Biologicals Supplier No. NBP272789AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRPS7 Monoclonal specifically detects MRPS7 in Human samples. It is validated for Western Blot.

Specifications

Antigen MRPS7
Applications Western Blot
Classification Monoclonal
Clone OTI5E11
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bMRP27a, bMRP-27a, mitochondrial ribosomal protein S7, MRP-S, MRP-S7,30S ribosomal protein S7 homolog, RPMS7,28S ribosomal protein S7, mitochondrial, RP-S7, S7mt
Gene Symbols MRPS7
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 38-242 of human MRPS7 (NP_057055) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51081.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.