Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRRF Antibody (7D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222564 Supplier Novus Biologicals Supplier No. NBP222564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRRF Monoclonal specifically detects MRRF in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MRRF
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:2000
Gene Accession No. NP_620132
Gene Alias mitochondrial, mitochondrial ribosome recycling factor, MTRRF
Gene Symbols MRRF
Host Species Mouse
Immunogen Recombinant human MRRF (56-262aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 92399
Test Specificity Anti-human MRRF mAb, clone 7D10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MRRF protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.