Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRRF Antibody (SR1378), Novus Biologicals™
SDP

Catalog No. NB387888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387888 50 μg
NB387889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387888 Supplier Novus Biologicals Supplier No. NBP32245150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MRRF Monoclonal antibody specifically detects MRRF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen MRRF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1378
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias mitochondrial, mitochondrial ribosome recycling factor, MTRRF
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRRF (Uniprot #: Q96E11)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.