Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRRF Antibody (SR1378), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | MRRF |
---|---|
Clone | SR1378 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
MRRF Monoclonal antibody specifically detects MRRF in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MRRF | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
mitochondrial, mitochondrial ribosome recycling factor, MTRRF | |
A synthesized peptide derived from human MRRF (Uniprot #: Q96E11) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1378 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
92399 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title