Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MRS 2179 tetrasodium salt
GREENER_CHOICE

Catalog No. p-4785146
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective P2Y1 antagonist

A competitive antagonist at P2Y1 receptors (KB = 100 nM). Selective over P2X1 (IC50 = 1.15 μM), P2X3 (IC50 = 12.9 μM), P2X2, P2X4, P2Y2, P2Y4 and P2Y6 receptors. Inhibits the upregulation of NTPDase1 by ATPγS.

Chemical Identifiers

CAS 1454889-37-2
Molecular Formula C11H17N5Na4O9P2

Specifications

Quantity 10 mg
Formula Weight 513.16
Purity >98%
Molecular Weight (g/mol) 517.19
Chemical Name or Material MRS 2179 tetrasodium salt
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.