Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRS 2179 tetrasodium salt, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
1454889-37-2
C11H17N5Na4O9P2
517.19
HLJPLHFSEAJJGC-ZKRIHRHSSA-N
2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
131848299
[(2R,3S,5R)-5-[6-(methylamino)purin-9-yl]-2-(phosphonooxymethyl)oxolan-3-yl] dihydrogen phosphate;sodium
CNC1=NC=NC2=C1N=CN2C3CC(C(O3)COP(=O)(O)O)OP(=O)(O)O.[Na].[Na].[Na].[Na]
1454889-37-2
C11H17N5Na4O9P2
517.19
HLJPLHFSEAJJGC-ZKRIHRHSSA-N
2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
131848299
[(2R,3S,5R)-5-[6-(methylamino)purin-9-yl]-2-(phosphonooxymethyl)oxolan-3-yl] dihydrogen phosphate;sodium
CNC1=NC=NC2=C1N=CN2C3CC(C(O3)COP(=O)(O)O)OP(=O)(O)O.[Na].[Na].[Na].[Na]

Selective P2Y1 antagonist

A competitive antagonist at P2Y1 receptors (KB = 100 nM). Selective over P2X1 (IC50 = 1.15 μM), P2X3 (IC50 = 12.9 μM), P2X2, P2X4, P2Y2, P2Y4 and P2Y6 receptors. Inhibits the upregulation of NTPDase1 by ATPγS.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 513.16
Percent Purity >98%
Chemical Name or Material MRS 2179 tetrasodium salt

Recommended Storage : Desiccate at -20°C