Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MRT 68921 dihydrochloride

Click to view available options
Quantity:
5 mg
25 mg
Description
MRT 68921 dihydrochloride is a potent ULK inhibitor (IC50 values are 1.1 and 2.9 nM for ULK2 and ULK1, respectively). Inhibits autophagy in mouse embryonic fibroblasts.
Specifications
Specifications
| Chemical Name or Material | N-[3-[[5-Cyclopropyl-2-[(1,2,3,4-tetrahydro-2-methyl-6-isoquinolinyl)amino]-4-pyrimidinyl]amino]propyl]cyclobutanecarboxamide dihydrochloride |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C25H34N6O.2HCl |
| Formula Weight | Observed MW: 507.5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction